Skip to Content
MilliporeSigma

52471

Fluorescein isothiocyanate–dextran

average mol wt 2,000,000

Synonym(s):

FITC–Dextran 2000, FITC–Dextran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 2,000,000

extent of labeling

0.001-0.020 mol FITC per mol glucose (substitution)

fluorescence

λex 493 nm; λem 520 nm in 0.1 M Tris pH 8.0

application(s)

cell analysis

Application

Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.
Fluorescein isothiocyanate–dextran (FITC–Dextran 2000) is use as a fluorescent probe to study the permeability of structures such as intercellular junctions involved in processes such as hearing and vascular permeability and the integrity of the blood brain barrier (BBB).


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library