Skip to Content
MilliporeSigma

52423

Dextran sulfate sodium salt from Leuconostoc spp.

low sulfate content, Mr ~40,000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Leuconostoc spp)

Quality Segment

form

powder

quality

low sulfate content

mol wt

Mr ~40,000

composition

Sulfur Content, 8-13%

loss

≤10% loss on drying

color

white to off-white

useful pH range

6.0-8 (10 g/L)

solubility

H2O: 100 mg/mL, clear, colorless to faintly greenish-yellow

storage temp.

room temp

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate increases binding of epithelial cells and lymphocytes to extracellular matrix proteins including fibronectin and laminin. In addition, dextran sulfate altered expreβion of cell surface receptors integrins.
Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Preparation Note

Sigma dextrans are derived from Leuconostoc mesenteroides.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library