Skip to Content
MilliporeSigma

Skip To

52044

Hexaethylene glycol monododecyl ether

BioXtra, ≥98.0% (TLC)

Synonym(s):

C12E6, Dodecylhexaglycol, Polyoxyethylene (6) lauryl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)6OH
CAS Number:
Molecular Weight:
450.65
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
221-282-3
Beilstein/REAXYS Number:
1715367
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

product line

BioXtra

assay

≥98.0% (TLC)

mol wt

450.65 g/mol

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C24H50O7/c1-2-3-4-5-6-7-8-9-10-11-13-26-15-17-28-19-21-30-23-24-31-22-20-29-18-16-27-14-12-25/h25H,2-24H2,1H3

InChI key

OJCFEGKCRWEVSN-UHFFFAOYSA-N

Other Notes

Non-ionic surfactant which effectively releases histamine from rat peritoneal mast cells[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7643774716P8925
assay

≥98.0% (TLC)

assay

≥98.0% (GC)

assay

≥98.0% (TLC)

assay

≥98% (GC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

mol wt

450.65 g/mol

mol wt

406.60 g/mol

mol wt

510.70 g/mol

mol wt

average mol wt 66,000

product line

BioXtra

product line

BioXtra

product line

BioXtra

product line

-

cation traces

Ca: ≤50 mg/kg, Co: ≤50 mg/kg, Cu: ≤50 mg/kg, K: ≤50 mg/kg, Mn: ≤50 mg/kg, Ni: ≤50 mg/kg, Zn: ≤50 mg/kg, Cr: ≤50 mg/kg, Mg: ≤50 mg/kg, Pb: ≤50 mg/kg, Cd: ≤50 mg/kg, Na: ≤50 mg/kg, Fe: ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

cation traces

-

description

non-ionic

description

non-ionic

description

non-ionic

description

non-ionic


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters