Skip to Content
MilliporeSigma
All Photos(1)

Documents

5162

Sigma-Aldrich

Bovine Collagen Type I

from bovine flexor tendon, powder, suitable for cell culture, used for 3D gel formation

Synonym(s):

Collagen Protein, Collagen Solution, Collagen Type I

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.75

product name

Collagen, from bovine flexor tendon

biological source

bovine flexor tendon

sterility

non-sterile

Assay

>96%

form

lyophilized powder (fibrous)

packaging

pkg of 1 g

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

Binding Specificity

Peptide Source: Elastin

Peptide Source: Fibronectin

Peptide Source: Laminin

shipped in

wet ice

storage temp.

room temp

Gene Information

bovine ... COL2A1(407142)

General description

This product is supplied as a lyophilized fibrous powder in a 1 gram package size. Bioburden and endotoxin levels are tested – this product is not considered sterile.

This collagen product is naturally cross-linked yielding a robust material for applications which require structure and strength. This product can be readily prepared into such forms as tissue scaffolds, foams, sponges, suspensions, coatings, putties, films and sheets but does not form hydrogels. Using typical cross-linking methods, this material can be tuned for optimal in vivo resorption. This collagen product is ideal for tissue engineering applications and uses with inorganic and biomaterials.

The product is provided in user-friendly packaging for use and storage. Avoid extended exposure to ambient environment since this material is hygroscopic.

Preparation Note

Type I bovine collagen, insoluble lyophilized fibrous powder, is extracted from bovine flexor tendon with the raw material sourced from closed/controlled herds of animals. This product is very difficult to dissolve, and after following the below protocol you will end up with a viscous solution.
Note: The following procedure is based on solubilization of 1 gram of collagen in 100 ml to initially prepare a 10 mg/ml collagen suspension. Smaller quantities and volumes may be used but the same ratios of collagen and solutions should be used.
1. Weigh out 1 gram of collagen fibrous powder.
2. Reconstitute 1 gram of collagen with 50 ml of cold purified water (50% of the final volume).
3. Stir the collagen with the water continuously mixing for a minimum of 15 minutes until the collagen is fully wetted and the suspension appears to be a semi-solution.
4. Add 50 ml of cold 0.02 M HCl to the collagen mixture and stir for a minimum of 10 minutes. This will yield a collagen concentration of 10 mg/ml with the suspension continuing to appear as a semi-solution.
5. Measure the pH - the mixture should have a pH of 2 to 3.
6. Using Waring stick blender or equivalent, homogenize the collagen mixture for a minimum of 15 minute at a high speed ensuring that the collagen is fully homogenized. Ensure that the temperature of mixture does exceed 24°C. Upon completion, there should be no visual solids in the viscous suspension. Air bubbles with be prevalent.
7. To remove air bubbles, stir the solution on a stir plate and pull a vacuum on the suspension. This will remove the air bubbles.
8. At this point if a collagen concentration of less than 10 mg/ml is desired, the collagen can be diluted with 0.01 M HCl.

Other Notes

Simultaneously vacuum filter 12 samples for rapid analysis of retentate or filtrate. Numbered positions on the filter plate assure accurate sample tracking. Sealing stoppers maintain vacuum on unused sample cups. Accessory extension barrels increase sample volumes from 15 to 50 mL.



Applications

General Filtration of 15 to 50 mL Samples, Preparation for Scintillation Counting



XX2702550 1225 Sampling Manifold



Replacement Parts

YY2214257 Hand Knob

5162 Top Plate

XX2702509 Filter O-ring, silicone

5163 Support Plate

XX2702510 Support Screen, 25 mm, polypropylene

XX11000PP Ball Valve, glass-filled polypropylene

XX2504755 Tubing, 1/4 in. x 23 in., latex



Accessories

XX2702552 Chamber O-rings and Plug Set

XX2702555 Sample Cup Extension Barrel

XX6200006P Filter Forceps, blunt end, stainless steel

XX7100004 Tubing 3/16 in. I.D. x 140 cm, silicone

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Parravicini et al.
PloS one, 15(4), e0231483-e0231483 (2020-04-23)
The GPR17 receptor, expressed on oligodendroglial precursors (OPCs, the myelin producing cells), has emerged as an attractive target for a pro-myelinating strategy in multiple sclerosis (MS). However, the proof-of-concept that selective GPR17 ligands actually exert protective activity in vivo is
Sylvain Merel et al.
Food chemistry, 289, 259-268 (2019-04-09)
Ethoxyquin (EQ) is an additive present in fish feed and its fate in fish should be carefully characterized due to food safety concerns regarding this compound. Therefore, the objective of this work was to identify the transformation products (TPs) of
Amjad Alagha et al.
Journal of biomaterials science. Polymer edition, 31(13), 1689-1705 (2020-05-14)
This study aims to achieve the principles of tissue engineering using biopolymers to be applied in the field of vital endodontic treatment to stimulate stem cells and engineering and regeneration of dentin tissue. the polymer blend was loaded with the
Yongjian Yue et al.
Journal of cellular biochemistry, 120(8), 12300-12310 (2019-02-28)
The disorders of hemostasis and coagulation were believed to be the main contributors to the pathogenesis of pulmonary thromboembolism (PTE), and platelets are the basic factors regulating hemostasis and coagulation and play important roles in the process of thrombosis. This
Paola Bonsi et al.
EMBO molecular medicine, 11(1) (2018-12-16)
Dopamine D2 receptor signaling is central for striatal function and movement, while abnormal activity is associated with neurological disorders including the severe early-onset DYT1 dystonia. Nevertheless, the mechanisms that regulate D2 receptor signaling in health and disease remain poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service