Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51536C

SAFC

RPMI-1640 Medium

with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

components

HEPES: 5958 mg/L
L-glutamine: 300 mg/L
NaHCO3: 2000 mg/L
phenol red: 5.310 mg/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium has been used to incubate dissected lung tissue.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Defining the inflammatory signature of human lung explant tissue in the presence and absence of glucocorticoid.
Tracy L R, et al.
F1000Research, 6 (2017)
Ananthakrishnan Soundaram Jeevarathinam et al.
Angewandte Chemie (International ed. in English) (2019-12-17)
We report a new approach to monitor drug release from nanocarriers via a paclitaxel-methylene blue conjugate (PTX-MB) with redox activity. This construct is in a photoacoustically silent reduced state inside poly(lactic-co-glycolic acid) (PLGA) nanoparticles (PTX-MB@PLGA NPs). During release, PTX-MB is
Ryoichi Arita et al.
Diabetes, 58(1), 215-226 (2008-10-09)
Leukocyte adhesion in retinal microvasuculature substantially contributes to diabetic retinopathy. Involvement of the Rho/Rho kinase (ROCK) pathway in diabetic microvasculopathy and therapeutic potential of fasudil, a selective ROCK inhibitor, are investigated. Localization of RhoA/ROCK and Rho activity were examined in
Tiffany M DeSimone et al.
International journal for parasitology, 39(4), 399-405 (2008-10-04)
Apicomplexan parasites employ multiple adhesive ligands for recognition and entry into host cells. The Duffy binding-like (DBL) and the reticulocyte binding protein-like (RBL) families are central to the invasion of erythrocytes by the malaria parasite. These type-1 transmembrane proteins are
Joke Patou et al.
Journal of inflammation (London, England), 6, 11-11 (2009-04-22)
The mast cell is a crucial effector cell in allergic rhinitis and other inflammatory diseases. During the acute allergic reaction preformed mediators such as histamine, but also de novo produced mediators such as leukotrienes (LTC4/D4/E4) and prostaglandins (PGD2) are released.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service