Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48130

Sigma-Aldrich

Kinetin

≥99.0% (HPLC)

Synonym(s):

6-Furfurylaminopurine, N6-Furfuryladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N5O
CAS Number:
Molecular Weight:
215.21
Beilstein:
21703
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99.0% (HPLC)

ign. residue

≤0.05%

mp

264-270 °C (dec.) (lit.)
269-271 °C (dec.)

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccco3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Phenolic profiling and bioactivity assessment of in vitro propagated Psidium cattleianum Sabine: This study highlights the use of kinetin in enhancing phenolic compound production in tissue cultures, demonstrating its potential in plant biotechnology and pharmaceutical applications (El-Deeb et al., 2024).

  • The impact of chemical and hormonal treatments to improve seed germination and seedling growth of Juniperus procera Hochst. ex Endi.: Research shows kinetin′s effectiveness in improving seed germination rates and seedling vigor, underlining its role as a valuable plant growth regulator (Jabbour and Alzahrani, 2024).

  • Callus Induction Followed by Regeneration and Hairy Root Induction in Common Buckwheat: Kinetin was used to successfully induce callus and facilitate regeneration in buckwheat, providing insights into its applications in genetic engineering and crop improvement (Omelchenko et al., 2024).

  • Long-term organogenic callus cultivation of Ranunculus illyricus L.: This study utilizes kinetin in the long-term cultivation of callus tissues, contributing to the conservation of plant species and enhancing our understanding of plant developmental biology (Kocot et al., 2024).

  • Optimizing medicinal hemp production with synergistic light-enhanced technologies and organic biorefinery approaches: Incorporating kinetin in light-enhanced growth protocols for hemp showcases its utility in optimizing plant production systems, potentially increasing yield and quality (Puttharak et al., 2024).


Biochem/physiol Actions

Kinetin has been studied as a treatment for many human disease states caused by pre-mRNA splicing errors, including familial dysautonomia and neurofibromatosis.

Other Notes

Plant growth regulator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Mohr et al.
Lehrbuch der Pflanzenphysiologie, 370-370 (1978)
C.O. Miller
Annu. Rev. Phytochem. Soc. Eur., 12, 395-395 (1961)
Eva Pros et al.
European journal of human genetics : EJHG, 18(5), 614-617 (2009-11-26)
Neurofibromatosis type 1 is one of the most common neurocutaneous autosomal dominant disorders. It is caused by mutations in the neurofibromatosis type 1 (NF1) gene and approximately 30-40% of them affect the correct splicing of NF1 pre-mRNA. In this report
Sergey Shabala et al.
Plant, cell & environment, 32(2), 194-207 (2008-11-22)
Nutrient acquisition in the mature root zone is under systemic control by the shoot and the root tip. In maize, exposure of the shoot to light induces short-term (within 1-2 min) effects on net K+ and H+ transport at the
Matthew M Hims et al.
Journal of molecular medicine (Berlin, Germany), 85(2), 149-161 (2007-01-09)
Mutations that affect the splicing of pre-mRNA are a major cause of human disease. Familial dysautonomia (FD) is a recessive neurodegenerative disease caused by a T to C transition at base pair 6 of IKBKAP intron 20. This mutation results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service