Skip to Content
MilliporeSigma

47036

Saponin

Molecular Biology, used as non-ionic surfactant

Synonym(s):

Glycosides, Saponins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
EC Number:
232-462-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

mol wt

1000-2000 g/mol

composition

Sapogenin, 8-25%

technique(s)

HPLC: suitable, protein quantification: suitable

CMC

0.001-0.01%

density

1.015-1.020 g/mL at 20 °C (5% in H2O)(lit.)

foreign activity

DNase, RNase, Protease, Phosphatase, none detected

SMILES string

C[C@]1(CCC23COC4([C@H]2C1)CC[C@@H]5[C@]6(CC[C@@H](C(C6CC[C@]5([C@@]4(C[C@H]3O)C)C)(C)C)O[C@H]7[C@@H]([C@H]([C@H](CO7)O[C@H]8[C@@H]([C@H]([C@@H](CO8)O)O)O)O[C@@H]9[C@@H]([C@H]([C@@H]([C@H](O9)CO)O)O)O)O[C@H]1[C@@H]([C@H]([C@@H]([C@H](O1)CO)O)O)O[C@@H]1[C@@H]([C@H]([C@@H]([C@H](O1)CO)O)O)O)C)C=O

InChI

1S/C58H94O27/c1-52(2)29-7-11-55(5)30(8-12-58-31-15-53(3,22-62)13-14-57(31,23-77-58)32(64)16-56(55,58)6)54(29,4)10-9-33(52)82-50-46(85-51-45(40(71)37(68)27(19-61)80-51)84-49-43(74)39(70)36(67)26(18-60)79-49)44(83-48-42(73)38(69)35(66)25(17-59)78-48)28(21-76-50)81-47-41(72)34(65)24(63)20-75-47/h22,24-51,59-61,63-74H,7-21,23H2,1-6H3/t24-,25-,26-,27-,28+,29?,30-,31+,32-,33+,34+,35-,36-,37-,38+,39+,40+,41-,42-,43-,44+,45-,46-,47+,48-,49-,50+,51+,53-,54+,55-,56+,57?,58?/m1/s1

InChI key

MAEBCGDGGATMSC-OSHGGGOQSA-N

General description

Saponins are a large family of amphiphilic glycosides of steroids and triterpenes. Saponins are found in many plants and are localized especially in the root, tuber, bark, leaves, seed, and fruit. They are also present in some animals, like sea cucumber.

Application

Glycoside used as a nonionic sufactant and adjuvant in vaccine development.
Saponin has been used:
  • in phosphate-buffered saline (PBS) to block cells for immunocytochemistry and confocal microscopy
  • to permeabilize cells for measuring total mitochondrial mass
  • as an internal control to extract saponin Holothuria nobilis Selenka

Biochem/physiol Actions

Saponins are nonionic surfactant that forms oil-in-water emulsions and acts as a protective colloid. They are used for the direct determination of metals in milk by flame atomic absorption spectrophotometry.
Saponins exhibit anti-tumor, antimicrobial, and anti-herbivore activity. They may also play a key role in plant defense. Saponins also possess several pharmaceutical properties, like anti-inflammatory, antifungal, antibacterial, anti-parasitic, and antiviral activities. Saponins can regulate rhizosphere bacteria in the soil.


Still not finding the right product?

Explore all of our products under Saponin


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library