Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46143

Supelco

Ofurace

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-(2,6-dimethylphenyl)-N-(tetrahydro-2-oxo-3-furanyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClNO3
CAS Number:
Molecular Weight:
281.73
Beilstein:
1653655
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cccc(C)c1N(C2CCOC2=O)C(=O)CCl

InChI

1S/C14H16ClNO3/c1-9-4-3-5-10(2)13(9)16(12(17)8-15)11-6-7-19-14(11)18/h3-5,11H,6-8H2,1-2H3

InChI key

OWDLFBLNMPCXSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Leslie et al.
Archives of environmental contamination and toxicology, 18(6), 876-880 (1989-11-01)
Ofurace, oxadixyl, and alachlor were studied for their effect on hepatic xenobiotic biotransformation in male rats by dosing i.p. with 1 or 100 mg/kg of chemical for 7 days. Ofurace decreased ethoxyresorufin-O-deethylase and aniline p-hydroxylase activities but induced ethoxycoumarin-O-deethylase activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service