Skip to Content
MilliporeSigma

46070

Ethyl cellulose

48.0-49.5% (w/w) ethoxyl basis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

autoignition temp.

698 °F

concentration

48.0-49.5 % (w/w) (ethoxyl)

color

white

refractive index

n20/D 1.47 (lit.)

viscosity

7-15 mPa.s, 6 % in toluene/ethanol 80:20(25 °C)

density

1.14 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

O1[C@H](C(C(C(C1COCC)OCC)OCC)OCC)O[C@@H]2C(OC(C(C2OCC)OCC)OCC)COCC

InChI

1S/C28H54O11/c1-9-29-17-19-21(31-11-3)23(32-12-4)26(35-15-7)28(38-19)39-22-20(18-30-10-2)37-27(36-16-8)25(34-14-6)24(22)33-13-5/h19-28H,9-18H2,1-8H3/t19?,20?,21?,22-,23?,24?,25?,26?,27?,28+/m1/s1

InChI key

MINRZDZESGXKDM-OLIDUAMRSA-N

Application

Ethyl cellulose is generated by converting a variable number of hydroxyl groups on the repeating glucose units to ethyl ether groups. Ethylcellulose is used in the preparation of biostructures such as matrices, microspheres, microparticles, pellets and tablets for use in drug delivery.

Preparation Note

Ethyl Cellulose is soluble in a wide range of solvents such as aliphatic alcohols, chlorinated solvents, and natural oils. It is insoluble in water, glycerin, and prolylene glycol.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Ethyl cellulose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library