Skip to Content
MilliporeSigma

Skip To

44612

Durcupan ACM

single component B, hardener 964

Synonym(s):

ACM resins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
13111001
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

General description

A water-soluble embedding medium for EM based on an aliphatic polyepoxide.

Application

Durcupan ACM, single component B, hardener 964 has been used for:
  • Embedding material for electron microscopy.
  • A method to prepare large resin sections for counting myelinated axons in rodent CNS and PNS structures.: This study presents a technique using Durcupan ACM for preparing large resin sections, enhancing the analysis of myelinated axons in rodent central and peripheral nervous systems.
  • Three-dimensional immersive virtual reality for studying cellular compartments in 3D models from EM preparations of neural tissues.: This research utilizes Durcupan ACM in electron microscopy to create 3D models of neural tissues, facilitating studies via immersive virtual reality.
  • Long-term effect of prazosin and losartan administration on blood pressure, heart, carotid artery, and acetylcholine induced dilation of cardiovascular system of young Wistar rats and SHR.: Explores the cardiovascular effects in rats using Durcupan ACM embedded samples for detailed tissue analysis.

Features and Benefits

Mixtures of Durcupan ACM present an interesting alternative to methacrylates for embedding sections for electron microscopy. They present the advantage that they harden uniformly, practically without shrinkage. All fixing agents normally used for electron microscopic work are also suitable for the Durcupan ACM embedding process. This process consists of three phases: dehydration of the tissue with acetone or ethyl alcohol, infiltration with the embedding agent, followed by hardening.

Legal Information

Durcupan is a trademark of Sigma-Aldrich Chemie GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L9774I475844611
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under Durcupan ACM


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters