Skip to Content
MilliporeSigma

Skip To

43944

DNA decontamination reagent

for removal of DNA on lab equipment

Synonym(s):

DNA AWAY

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.52
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

liquid

storage temp.

room temp

General description

DNA Decontamination Reagent is a ready-to-use surfactant that removes DNA from lab equipment. Suitable for PCR equipment, glassware, plastic, and stainless steel lab surfaces.

Application

DNA decontamination reagent has been used to remove DNA from laminar flow cabinet before bacterial artificial chromosome- array comparative genomic hybridization (BAC aCGH) and single nucleotide polymorphism (SNP) array protocols.

Other Notes

Do not use DNA Decontamination Reagent on aluminium, soft metal or gaskets and seals. This product is not intended as reactant. Do not add to reaction.

Disclaimer

Gloves should be worn when handling this product. DNA Decontamination Reagent is alkaline in nature and will cause irritation if prolonged contact with the skin is allowed. In case of contact with eyes, immediately flush with water for fifteen minutes and contact a physician. If swallowed, Do not induce vomiting. Give plenty of water and contact a physician immediately.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.57010PNS10103097S
storage temp.

room temp

storage temp.

-

storage temp.

2-8°C

storage temp.

-

grade

Molecular Biology

grade

-

grade

-

grade

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

-

form

liquid

form

liquid


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Does this solution also removes DNases from lab equipment?

    1 answer
    1. This product, when used properly, will effectively remove DNA and DNase contamination from plastic, stainless steel, and glass laboratory surfaces.

      Helpful?

Reviews

No rating value

Active Filters