Skip to Content
MilliporeSigma
All Photos(1)

Documents

4374771

Sigma-Aldrich

mTRAQ® Reagent 10 Assay Kit

Synonym(s):

Δ8 mTRAQ Reagent Pack, 50 Unit mTRAQ Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.24

Quality Level

storage temp.

−20°C

General description

Contains mTRAQ® Reagents Δ0, Δ4, and Δ8, sufficient for 10 mTRAQ assays. One unit of reagent can label up to 100 μg of protein. The kit has 10 tubes each of Δ0, Δ4, and Δ8 reagents, each containing 1 unit of reagent, and the Protocol and Quick Reference Card. Optional Multiplex Buffer kit (Cat. No. 4381664) is to be ordered separately.

Analysis Note

To view the Protocol for use of mTRAQ Reagents, please visit this Protocol link.

To view the Quick Reference Card for use of mTRAQ Reagents, please visit this Quick Reference Card link.

Legal Information

MTraq is a registered trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service