Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43394

Sigma-Aldrich

Anti-Mouse IgG−Atto 550 antibody produced in goat

1 mg/mL protein

Synonym(s):

Atto 550-Anti-Mouse-IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

Atto 550 conjugate

Quality Level

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

50% glycerol as stabilizer

species reactivity

mouse

concentration

1 mg/mL protein

technique(s)

immunofluorescence: suitable
protein array: 2.8 μg/mL

fluorescence

λex 550 nm; λem 576 nm in PBS

storage temp.

−20°C

target post-translational modification

unmodified

Physical form

Atto 550 goat anti-mouse IgG (whole molecule) is provided in unit sizes of 1 ml as 1 mg/ml solutions in 0.1 M sodium phosphate, 0.1 M NaCl, pH 7.5, containing 5 mM sodium azide as a preservative.

Analysis Note

unconjugated dye ≤5% of total fluorescence

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piyush Mishra et al.
Biochemistry and biophysics reports, 11, 138-146 (2017-09-29)
Macroautophagy is a cellular response to starvation wherein superfluous and damaged cytoplasmic constituents are degraded to provide energy for survival and to maintain cellular homeostasis. Dysfunctional autophagy is attributed to disease progression in several pathological conditions and therefore, autophagy has
Juan Eduardo Rodriguez-Gatica et al.
Development (Cambridge, England), 149(20) (2022-07-29)
Organoids are stem cell-derived three-dimensional cultures offering a new avenue to model human development and disease. Brain organoids allow the study of various aspects of human brain development in the finest details in vitro in a tissue-like context. However, spatial
Albino Bacolla et al.
Nucleic acids research, 49(1), 221-243 (2020-12-11)
Human genome stability requires efficient repair of oxidized bases, which is initiated via damage recognition and excision by NEIL1 and other base excision repair (BER) pathway DNA glycosylases (DGs). However, the biological mechanisms underlying detection of damaged bases among the
Jean Defourny et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(16), 8010-8017 (2019-04-03)
Noise overexposure causes oxidative stress, leading to auditory hair cell damage. Adaptive peroxisome proliferation involving pejvakin, a peroxisome-associated protein from the gasdermin family, has been shown to protect against this harmful oxidative stress. However, the role of pejvakin in peroxisome
Baptiste Libé-Philippot et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(30), 7765-7774 (2017-07-15)
Many genetic forms of congenital deafness affect the sound reception antenna of cochlear sensory cells, the hair bundle. The resulting sensory deprivation jeopardizes auditory cortex (AC) maturation. Early prosthetic intervention should revive this process. Nevertheless, this view assumes that no

Articles

Immunoblotting (Western blot transfer) is a common technique in modern proteomics research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service