Skip to Content
MilliporeSigma

392M-1

Langerin (12D6) Mouse Monoclonal Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
12D6, monoclonal
Application:
IHC (p)
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

12D6, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (392M-14), vial of 0.5 mL concentrate (392M-15), bottle of 1.0 mL predilute (392M-17), vial of 1.0 mL concentrate (392M-16), bottle of 7.0 mL predilute (392M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2bκ

control

Langerhans cell histiocytosis

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... CD207(50489)

General description

Langerin is a type II transmembrane C-type lectin associated with the formation of Birbeck granules in Langerhans cells. The demonstration of langerin immunoreactivity is considered an adjunct or alternative to CD1a antigen expression as evidence to aid in the diagnosis of Langerhans cell histiocytosis. Evaluation of langerin expression is valuable in circumstances where a diagnosis of Langerhans cell histiocytosis is suspected, but cannot be confirmed due to lack of CD1a immunoreactivity. A panel of antibodies against CD1a, langerin, CD21, CD23, CD35 and S100 is very useful in the distinction of Langerhans cell histiocytosis, histiocytic sarcoma, interdigitating dendritic cell sarcoma, follicular dendritic cell sarcoma, disseminated juvenile xanthogranuloma, and Rosai-Dorfman disease (sinus histiocytosis with massive lymphadenopathy).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Analysis Note


IVD

IVD

IVD

RUO

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]
Langerin Positive Control Slides, Product No. 392S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
392M-140841683140528
392M-160841683140542
392M-150841683140535
392M-170841683140559
392M-180841683140566