Skip to Content
MilliporeSigma

391A

Leukocyte Peroxidase (Myeloperoxidase) Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

DAB substrate kit

Quality Segment

usage

sufficient for 10-120 manual assays

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

dilution

(for histology)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

General description

Used to demonstrate polymorphonuclear cells in blood or bone marrow films. Each kit has sufficient reagents for 10 uses.

Application

Diaminobenzidine, as a benzidine substitute for myeloperoxidase, has less tinctorial power than benzidine. This made DAB much less attractive to histochemists. Hanker and associates improved DAB methodology. According to their modification, the brown reaction product is first intensified with copper salts and followed by application of Gill′s modified Papanicolaou stain, resulting in intense grey-black staining at sites of neutrophil and monocyte peroxidase.
The Leukocyte Peroxidase (Myeloperoxidase) kit is intended for use as a general laboratory histochemical stain for the demonstration of leukocyte peroxidase. This manual, qualitative procedure identifies leukocyte peroxidase in blood or bone marrow films of human specimens.


Still not finding the right product?


Kit Components Only

Product No.
Description

  • Diaminobenzidine 10 vial(s)

  • Copper nitrate 2 vial(s)

  • Gill Modified EA solution 100 mL

  • Glutaraldehyde solution 2 x 75

  • Hematoxylin Solution, Gill No. 3 100 mL

  • Scott's Tap Water Substitute Concentrate 100 mL

  • TRIZMAL™ Buffer Concentrate 50 mL

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Ox. Liq. 1 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Kidney, Respiratory system

supp_hazards

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk

WGK 3

flash_point_f

57.2 °F

flash_point_c

14 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library