Skip to Content
MilliporeSigma

3872

Fast Garnet GBC Base Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
41116124
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

7.0 mg/mL

pH

<1.0

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Cc1ccccc1\N=N\c2ccc(N)c(C)c2

InChI

1S/C14H15N3/c1-10-5-3-4-6-14(10)17-16-12-7-8-13(15)11(2)9-12/h3-9H,15H2,1-2H3/b17-16+

InChI key

PFRYFZZSECNQOL-WUKNDPDISA-N

Application

Intended for use in Sigma Procedure 387 for the demonstration of Acid Phosphatase (TRAP).

Other Notes

Fast Garnet GBC base, 7.0 mg/mL, in 0.4 mol/L hydrochloric acid with stabilizer


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Met. Corr. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library