Skip to Content
MilliporeSigma

Skip To

37442

Dihydroxyacetone phosphate lithium salt

≥95.0% (TLC)

Synonym(s):

1-Hydroxy-3-(phosphonooxy)-2-propanone lithium salt, DHAP, Glycerone phosphate lithium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H7O6P · xLi+
CAS Number:
Molecular Weight:
170.06 (free acid basis)
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (TLC)

form

powder

storage temp.

−20°C

SMILES string

OCC(COP(O)(O)=O)=O

InChI

1S/C3H7O6P/c4-1-3(5)2-9-10(6,7)8/h4H,1-2H2,(H2,6,7,8)

InChI key

GNGACRATGGDKBX-UHFFFAOYSA-N

Biochem/physiol Actions

Dihydroxyacetone phosphate (DHAP) is a metabolic intermediate involved in many pathways, including glycolysis, gluconeogenesis, glycerol metabolism, phosphatidic acid synthesis, fat metabolism, and the Calvin cycle.

Analysis Note

may contain up to 2-mol-equivalents water

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
788327915692034
description

≥95.0% (TLC)

description

≥90% (TLC)

description

≥95% (TLC)

description

≥95.0% (TLC)

assay

≥95.0% (TLC)

assay

≥90% (TLC)

assay

≥95% (TLC)

assay

≥95.0% (TLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

phosphatidic acid synthesis reagent DHAP

-

-

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters