Skip to Content
MilliporeSigma

36617

Diphenyl phthalate

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4-1,2-(CO2C6H5)2
CAS Number:
Molecular Weight:
318.32
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
201-546-4
Beilstein/REAXYS Number:
2473390
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

mp

74-76 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

O=C(Oc1ccccc1)c2ccccc2C(=O)Oc3ccccc3

InChI

1S/C20H14O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h1-14H

InChI key

DWNAQMUDCDVSLT-UHFFFAOYSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Diphenyl phthalate


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library