Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358M-1

Sigma-Aldrich

MUM1 (MRQ-8) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-8, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (358M-14)
vial of 0.5 mL concentrate (358M-15)
bottle of 1.0 mL predilute (358M-17)
vial of 1.0 mL concentrate (358M-16)
bottle of 7.0 mL predilute (358M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

B-cell chronic lymphocytic leukemia (B-CLL), plasma cell myeloma, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, nuclear

Gene Information

human ... PWWP3A(84939)

General description

MUM1 (multiple myeloma oncogene-1)/IRF4 (interferon regulatory factor 4) is a 50 kDa protein encoded by MUM1 gene, and a member of the interferon regulatory factor family of transcription factors. MUM1/IRF4 is expressed in the nuclei of plasma cells and a small percentage of germinal center (GC) B-cells located in the “light zone”. This antibody stains MUM1 protein, which is expressed in a subset of B-cells in the light zone of the germinal center, plasma cells, activated T-cells, and a wide spectrum of related hematolymphoid neoplasms derived from these cells. Therefore, this antibody is useful for the subclassiffication of lymphoid malignancies and an excellent marker for Reed-Sternberg cells of classic Hodgkin disease in combination with anti-CD30.

Quality


IVD

IVD

IVD

RUO

Linkage

MUM1 Positive Control Slides, Product No. 358S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gaidano et al.
Leukemia, 14(4), 563-566 (2000-04-14)
In recent times, the field of B cell lymphoma histogenesis has progressed rapidly due to the increasing availability of histogenetic markers. Genotypic markers of B cell histogenesis are represented by mutations of IgV and BCL-6 genes, which are somatically acquired
Antonino Carbone et al.
British journal of haematology, 117(2), 366-372 (2002-04-26)
Biological and clinical studies have shown that Hodgkin's disease (HD) can be divided into two major categories, termed nodular lymphocyte predominance HD (NLP HD) and classic HD (CHD). Within CHD four subtypes have been distinguished: nodular sclerosis, mixed cellularity, lymphocyte
Y Natkunam et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 14(7), 686-694 (2001-07-17)
The gene encoding MUM1 was characterized as a possible translocation partner in chromosomal abnormalities involving a significant number of multiple myelomas. The overexpression of the MUM1 protein as a result of translocation t(6;14) (p25;q32) identified MUM1 as a putative regulatory
Mathewos Tessema et al.
PloS one, 7(4), e34850-e34850 (2012-04-13)
Aberrant cytosine methylation affects regulation of hundreds of genes during cancer development. In this study, a novel aberrantly hypermethylated CpG island in cancer was discovered within the TOX2 promoter. TOX2 was unmethylated in normal cells but 28% lung (n =
B Falini et al.
Blood, 95(6), 2084-2092 (2000-03-09)
A new monoclonal antibody (MUM1p) was used to study the cell/tissue expression of human MUM1/IRF4 protein, the product of the homologous gene involved in the myeloma-associated t(6;14) (p25;q32). MUM1 was expressed in the nuclei and cytoplasm of plasma cells and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service