Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

352R-1

Sigma-Aldrich

Napsin A (EP205) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP205, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (352R-14)
vial of 0.5 mL concentrate (352R-15)
bottle of 1.0 mL predilute (352R-17)
vial of 1.0 mL concentrate (352R-16)
bottle of 7.0 mL predilute (352R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

lung adenocarcinoma, renal cell carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... NAPSA(9476)

General description

Napsin is a pepsin-like aspartic proteinase, in the A1 clan of the AA clade of proteinases. There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein with the molecular weight of approximately 38 kDa. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.1-5

Quality


IVD

IVD

IVD

RUO

Linkage

Napsin A Positive Control Slides, Product No. 352S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annika Dejmek et al.
Diagnostic cytopathology, 35(8), 493-497 (2007-07-20)
The purpose of this study was to test napsin A as a diagnostic marker of metastatic lung adenocarcinoma in pleural effusions, and to compare its performance with TTF-1. Napsin A and TTF-1 reactivities were determined immunohistochemically on formalin-fixed paraffin embedded
Kentaro Inamura et al.
The American journal of surgical pathology, 29(5), 660-665 (2005-04-16)
Primary pulmonary adenocarcinomas with enteric differentiation (PAED) are mainly composed of tall-columnar cells that show similarity to intestinal epithelia and colorectal carcinomas. In this study, we analyzed the immunostaining profiles of 7 PAEDs in comparison with 14 metastatic colorectal carcinomas
Jaishree Jagirdar
Archives of pathology & laboratory medicine, 132(3), 384-396 (2008-03-06)
Immunohistochemistry is a very valuable and often used tool in the differential diagnosis of lung carcinomas whether primary or secondary to the lung. The most useful application is in distinguishing primary lung tumors from metastatic tumors to the lung from
Justin A Bishop et al.
Human pathology, 41(1), 20-25 (2009-09-11)
Recent advances in the treatment of pulmonary adenocarcinoma have increased the need for accurate typing of non-small cell carcinomas. Immunohistochemistry for thyroid transcription factor-1 is widely used in the diagnosis of pulmonary adenocarcinomas because it marks approximately 75% of lung
Jiqing Ye et al.
Applied immunohistochemistry & molecular morphology : AIMM, 19(4), 313-317 (2011-04-06)
Differentiation of primary from metastatic adenocarcinoma in the lung can be challenging, and it demands sensitive and specific biomarkers, especially when the tissue for diagnosis is limited. Thyroid transcription factor-1 (TTF-1) has been considered a reliable marker for adenocarcinoma of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service