Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337M-8

Sigma-Aldrich

TAG-72 (B72.3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

B72.3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (337M-84)
vial of 0.5 mL concentrate (337M-85)
bottle of 1.0 mL predilute (337M-87)
vial of 1.0 mL concentrate (337M-86)
bottle of 7.0 mL predilute (337M-88)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

lung adenocarcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-B72.3 recognizes a high molecular weight glycoprotein called TAG-72 that is present in human adenocarcinomas and in lesser amounts, non-neoplastic tissues. It has also been found to be useful to distinguish between mesothelioma and adenocarcinoma, however, false positive reactions can occur so results must be interpreted with the utmost caution.

Quality


IVD

IVD

IVD

RUO

Linkage

TAG-72 Positive Control Slides, Product No. 337S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K G Osteen et al.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists, 11(3), 216-220 (1992-07-01)
Compared with the degree of investigation of stage-specific expression of tumor-associated glycoprotein-72 (TAG-72) in fundal endometrium, other regions of the female reproductive tract have not received comparable attention. Regional, cell-type-specific, and temporal differences in estrogen receptor and progesterone receptor distribution
T S Kline et al.
Cancer, 63(11), 2253-2256 (1989-06-01)
Monoclonal antibody B72.3 (MoAb B72.3) is a potentially valuable diagnostic adjunct when applied to aspiration biopsy cytology (ABC). In this prospective study, its reactivity was tested on the ABC from 25 breast lesions interpreted as suspicious. The stain was applied
W W Johnston et al.
Human pathology, 17(5), 501-513 (1986-05-01)
Monoclonal antibody B72.3 has been shown to be reactive with a high-molecular-weight glycoprotein complex termed TAG(tumor-associated glycoprotein)-72. By the avidin-biotin immunoperoxidase method, fine needle aspirates and corresponding surgically excised tumor tissues from both malignant and benign tissues were analyzed for
David C Chhieng et al.
Human pathology, 34(10), 1016-1021 (2003-11-11)
Ovarian carcinoma has a high mortality rate, because most ovarian carcinomas are detected at a late stage. Traditional therapies, such as surgical debulking and chemotherapy, have not been successful in improving the long-term survival of these patients. Alternative therapies targeting
J Lundy et al.
Annals of surgery, 203(4), 399-402 (1986-04-01)
Fine needle aspirations of breast masses were evaluated to establish a cytologic diagnosis. In 45 cases, the sensitivity was 73%, specificity 100%, and diagnostic accuracy 84%. Fifty cases (27 malignant and 23 benign) were evaluated by immunoperoxidase staining of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service