Skip to Content
MilliporeSigma

Skip To

31388

Dextran from Leuconostoc spp.

Mr ~6,000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
EC Number:
232-677-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Leuconostoc spp)

Quality Segment

form

powder

mol wt

Mr ~6,000

color

white

mp

483  °C ((901 °F ))

solubility

water: 1.5 g/10 mL, colorless to faintly yellow

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

General description

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components. Low molecular weight dextrans are often preferred versus high molecular weight dextrans due to their different viscosities, aggregation and permeation properties.
Dextran is an extracellular bacterial polymer of D-glucopyranose. It mostly comprises α-(1→6) linkage in the main chain and a variable number of α-(1→2), α-(1→3), α-(1→4) branched linkages.

Application

Dextran from Leuconostoc spp. has been used:
  • for the glycation of caseinate to investigate the influence of an increase in carbohydrate chain length compared to glucose
  • for the mass calibration of matrix-assisted laser desorption ionization-time of flight mass spectroscopy (MALDI-TOF) for the characterization of silver nanoparticles (AgNPs)
  • as a non-toxic stabilizer to assess the in vivo administration of tungsten trioxide(WO3) nanoparticles

Biochem/physiol Actions

Dextran is used as a drug, particularly as a blood plasma volume expander. It is also useful as an adjuvant, emulsifier, carrier, and stabilizer in the food, pharmaceutical, and chemical industries. Sephadex is a cross-linked dextran that is commonly used for the separation and purification of protein. In the food industry, dextran is applicable as a thickener for jam and ice cream. It hinders the crystallization of sugar and augments moisture retention. This preserves the flavor and appearance of several food items.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
313920918431398
biological source

(Leuconostoc spp)

biological source

(Leuconostoc spp)

biological source

(Leuconostoc spp.)

biological source

(leuconostoc mesenteroides)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

water: 1.5 g/10 mL, colorless to faintly yellow

solubility

water: 1.5 g/10 mL, clear, colorless

solubility

water: 1.5 g/10 mL, clear to slightly hazy, colorless to faintly yellow

solubility

-

form

powder

form

powder

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

color

white

color

white

color

-

color

white


Still not finding the right product?

Explore all of our products under Dextran from Leuconostoc spp.


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters