Skip to Content
MilliporeSigma

3023

AptaFluor® SAH Methyltransferase Assay

Synonym(s):

Methyltransferase Activity Assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41122100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

storage temp.

−70°C

General description

The AptaFluor® SAH Methyltransferase TR-FRET Assay is a universal assay for enzymes that convert S-adenosylmethionine (SAM) to S-adenosylhomocysteine (SAH), with a far-red, time-resolved Förster-resonance-energy-transfer (TR-FRET) readout. The assay can be used for all enzymes within the histone methyltransferase (HMT) and DNA methyltransferase (DNMT) families. The assay is designed specifically for high-throughput screening (HTS), with a simple mix-and-read format and 2 liquid addition steps. It is run in an endpoint mode: methyltransferase (MT) enzyme reactions are quenched with a universal stop reagent (provided) and then the detection reagents are added.

The assay offers reagent stability and compatibility with commonly used multimode plate readers. The AptaFluor® SAH Methyltransferase TR-FRET Assay provides the following benefits:
  • Simple, mix-and-read format ideal for HTS.
  • Robust MT detection under initial velocity conditions (=20% conversion of SAM to SAH).
  • Accommodates SAM concentrations as low as 100 nM and as high as 5 μM.
  • Far-red tracer further minimizes interference from fluorescent compounds and light scattering.

View full Transcreener® product list

Physical form

Kit with buffered aqueous solutions

Preparation Note

3023-B = 100 assay, 96-well
3023-1K = 1,000 assay, 384-well

Legal Information

AptaFluor is a registered trademark of BellBrook Labs
Transcreener is a registered trademark of BellBrook Labs




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



L Latini et al.
Cell death & disease, 5, e1404-e1404 (2014-09-05)
Spinal cord injury (SCI) is a devastating condition of CNS that often results in severe functional impairments for which there are no restorative therapies. As in other CNS injuries, in addition to the effects that are related to the primary
Kalyan C Kondapalli et al.
The Journal of biological chemistry, 287(43), 36239-36250 (2012-09-06)
Human NHA2, a newly discovered cation proton antiporter, is implicated in essential hypertension by gene linkage analysis. We show that NHA2 mediates phloretin-sensitive Na(+)-Li(+) counter-transport (SLC) activity, an established marker for hypertension. In contrast to bacteria and fungi where H(+)
Aileen K Sewell et al.
iScience, 25(5), 104186-104186 (2022-04-29)
The protein kinase complex target of rapamycin complex 1 (TORC1) is a critical mediator of nutrient sensing that has been widely studied in cultured cells and yeast, yet our understanding of the regulatory activities of TORC1 in the context of



Global Trade Item Number

SKUGTIN
229164-5G04061832339054
229164-25G04061832339047
3023-B04061835357444
3023-1K04061835357116