Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.84
UNSPSC Code:
41122100
shipped in
dry ice
storage temp.
−20°C
General description
The Transcreener® UDP2 TR-FRET Red Assay is a competitive immunoassay for UDP with a far-red, time-resolved Förster-resonance-energy-transfer (TR-FRET) readout. Because it is highly selective for UDP, the assay can be used with any enzyme that generates UDP from a UDP-sugar donor. Examples include glycosyltransferase, galactosyltransferase, glucuronyltransferase, N-acetylglucosamyltransferase, N-acetylgalactosyltransferase, xylosyltransferase, and glycogen, cellulose, lactose, and hyaluronan synthases. The Transcreener® assay is designed specifically for high-throughput screening (HTS), with a single-addition, mix-and-read format. It offers reagent stability and compatibility with commonly used multimode plate readers. The generic nature of the Transcreener® HTS assay platform eliminates delays involved in assay development for new HTS targets and greatly simplifies compound and inhibitor profiling across multiple target families.
The Transcreener® UDP2 TR-FRET Red Assay provides the following benefits:
View full Transcreener® product list
The Transcreener® UDP2 TR-FRET Red Assay provides the following benefits:
- Accommodates UDP-sugar donor concentrations ranging from 1 μM to 1,000 μM.
- Excellent data quality (Z′ = 0.7) at low substrate conversion (typically 10–30%).
- Overcomes the need for time-consuming, one-off assay development for individual members within an enzyme family by using a single set of assay reagents that detect an invariant product.
- Time-gated detection method largely eliminates interference that can result from prompt fluorescence of test compounds.
- Far-red tracer further minimizes interference from fluorescent compounds and light scattering.
View full Transcreener® product list
Physical form
Kit with buffered aqueous solutions
Preparation Note
3022-1K = 1,000 assay, 384-well
3022-10K = 10,000 assay, 384-well
3022-10K = 10,000 assay, 384-well
Legal Information
Transcreener is a registered trademark of BellBrook Labs
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Drake Phelps et al.
Scientific reports, 7(1), 11244-11244 (2017-09-13)
Changes in resident microbiota may have wide-ranging effects on human health. We investigated whether early life microbial disruption alters neurodevelopment and behavior in larval zebrafish. Conventionally colonized, axenic, and axenic larvae colonized at 1 day post fertilization (dpf) were evaluated using
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 424633-5G | 04061832103778 |
| 424633-25G | 04061832103761 |
| 8201180025 | 04022536452395 |
| 8201180100 | 04022536452401 |
| 3022-1K | 04061835356980 |
| 3022-10K | 04061835357109 |