Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

271M-1

Sigma-Aldrich

Inhibin, alpha (R1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

R1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (271M-14)
vial of 0.5 mL concentrate (271M-15)
bottle of 1.0 mL predilute (271M-17)
vial of 1.0 mL concentrate (271M-16)
bottle of 7.0 mL predilute (271M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:5-1:100

isotype

IgG2a

control

adrenal cortex

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... INHA(3623)

General description

Anti–inhibin alpha is an antibody against a peptide hormone which has demonstrated utility in the differentiation between adrenal cortical tumors and renal cell carcinoma. Sex cord stromal tumors of the ovary as well as trophoblastc tumors also demonstrate cytoplasmic positivity with this antibody. This antibody has been used to make the differential diagnosis of intrauterine vs. ectopic pregnancy in endometrial curettings.

Quality


IVD

IVD

IVD

RUO

Linkage

Inhibin, alpha Positive Control Slides, Product No. 271S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonardo D Santos et al.
Pathology, 35(3), 217-223 (2003-09-26)
Using archival material, we studied the immunoreactivity and utility of monoclonal anti-human inhibin alpha subunit in the identification of chorionic villi (CV) and trophoblastic subpopulations in endometrial curettings (EC) from patients who had intra-uterine, ectopic, molar and, particularly, probable intra-uterine
X Matias-Guiu et al.
Human pathology, 29(8), 840-845 (1998-08-26)
Müllerian inhibiting substance (MIS), alpha-inhibin, and CD99 are expressed by normal and neoplastic sex cord and stromal gonadal derivatives. The expression of MIS, inhibin, and CD99 was evaluated by immunohistochemistry in a series of 19 sex cord-stromal tumors and seven
T J Pelkey et al.
Human pathology, 30(1), 26-31 (1999-01-29)
Placental trophoblasts are the primary source of serum inhibin during pregnancy. We sought to characterize inhibin immunolabeling in trophoblastic neoplasms and compare the results with those for established markers of trophoblastic differentiation. Formalin-fixed, paraffin-embedded tissues from three normal term placentas
K Yamashita et al.
American journal of obstetrics and gynecology, 177(6), 1450-1457 (1998-01-10)
Our purpose was to examine the cellular localization of inhibin subunits and messenger ribonucleic acid expressions for the inhibin subunits and the serum levels of inhibin A and inhibin B in human ovarian sex cord stromal tumors. We examined the
W G McCluggage
Histopathology, 40(4), 309-326 (2002-04-12)
Recent years have witnessed significant developments in the use of immunohistochemistry in diagnostic gynaecological pathology. This review details the most significant of these. In ovarian pathology, differential cytokeratin staining (CK7 and 20) assists in distinguishing between a primary ovarian adenocarcinoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service