Skip to Content
MilliporeSigma

Skip To

27029

Sodium cholate hydrate

≥97.0% (dried material, NT)

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H39NaO5 · xH2O
CAS Number:
Molecular Weight:
430.55 (anhydrous basis)
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
206-643-5
Beilstein/REAXYS Number:
3582354
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

anionic

Quality Segment

assay

≥97.0% (dried material, NT)

form

powder

optical activity

[α]/D 26.0 to 34.0°°, c = 2% (solution in purified water, CALCULATED FOR DRIED SUBSTANCE)

mol wt

micellar avg mol wt 900-1300

aggregation number

2-3

loss

≤5% loss on drying

CMC

9-15 mM (20-25°C)

solubility

H2O: 0.1 M, H2O: 100 mg/mL

HLB

18

SMILES string

O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3[C@H](O)CC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C

InChI

1S/C24H40O5.Na.H2O/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;;/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29);;1H2/q;+1;/p-1/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-;;/m1../s1

InChI key

MUVVIYFKOVLQHL-RCVKHMDESA-M

General description

Sodium cholate hydrate is a bio-surfactant which can be used as a reducing reagent in aqueous solution.
Sodium cholate hydrate is a non-denaturing, anionic detergent.

Application


  • Optimization in Transdermal Drug Delivery: Applied in the formulation of ultra-elastic nanovesicles, sodium cholate hydrate enhances the bioavailability and anti-inflammatory efficacy of deflazacort, demonstrating its potential in transdermal drug delivery systems, which can be adapted for diagnostic purposes (Ali et al., 2021).

  • Nanocarrier Formulations for IVDs: The study utilizes sodium cholate hydrate in the surface modification of bilosomes, serving as a functional and biocompatible nanocarrier for hybrid compounds, a method applicable to the formulation of innovative IVD reagents (Waglewska et al., 2020).

  • IVD Reagent Formulation for Skin Delivery: Sodium cholate hydrate is featured in the development of nano-transfersomes for the delivery of antioxidants, showcasing its role in creating skin-applied diagnostic reagents that offer antioxidant and anti-aging effects (Avadhani et al., 2017).

  • Sorting and Separation Techniques in Nanotechnology: Demonstrating the affinity-mediated sorting order reversal, sodium cholate hydrate is critical in density gradient ultracentrifugation techniques, highlighting its utility in the precise sorting and preparation of materials for diagnostic assays (Jang et al., 2016).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C6445C1254C9282
assay

≥97.0% (dried material, NT)

assay

≥99%

assay

≥99%

assay

≥99.0% (HPLC)

form

powder

form

powder

form

powder

form

powder

aggregation number

2-3

aggregation number

2-3

aggregation number

2-3

aggregation number

2-3

CMC

9-15 mM (20-25°C)

CMC

9-15 mM (20-25°C)

CMC

9-15 mM (20-25°C)

CMC

9-15 mM (20-25°C)

HLB

18

HLB

18

HLB

18

HLB

18

description

anionic

description

anionic

description

anionic

description

anionic


Still not finding the right product?

Explore all of our products under Sodium cholate hydrate


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters