Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24533C

SAFC

EX-CELL® EBx PRO-II Serum-Free Media

Animal-component free, Chemically defined, dry powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

EX-CELL® EBx Serum-Free Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

Quality Level

description

Animal-component-free, low protein
for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C
2-8°C

Looking for similar products? Visit Product Comparison Guide

Quantity

Formulated to contain 12.4 grams of powder per liter of medium.

Legal Information

EBx is a trademark of Vivalis SA
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service