Skip to Content
MilliporeSigma

2108

E-Toxate Reagent

Single test vial

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.83
UNSPSC Code:
12352204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

sensitivity measuring range

0.05-0.1 EU/mL

storage temp.

−20°C

Other Notes

10 mL of Catalog No. 2107 ("Endotoxin-free" water) is supplied with each 1-vial and 10-vial order. 2 × 10 mL is supplied with 100-vial orders.
Ready-to-use single test vials containing E-TOXATE reagent. Pipette test solution directly into the vial.

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Tanguy Lechertier et al.
Nature communications, 11(1), 2810-2810 (2020-06-06)
The overexpression of the protein tyrosine kinase, Focal adhesion kinase (FAK), in endothelial cells has implicated its requirement in angiogenesis and tumour growth, but how pericyte FAK regulates tumour angiogenesis is unknown. We show that pericyte FAK regulates tumour growth
Tibor Sághy et al.
Cell death & disease, 10(6), 439-439 (2019-06-06)
Transglutaminase 2 (TG2) is a multifunctional protein that promotes clearance of apoptotic cells (efferocytosis) acting as integrin β3 coreceptor. Accumulating evidence indicates that defective efferocytosis contributes to the development of chronic inflammatory diseases. Obesity is characterized by the accumulation of



Global Trade Item Number

SKUGTIN
119474-5G04061832097770
119474-25G04061832097763
2108-10VL04061833291382