Skip to Content
MilliporeSigma

181B

Lymphocyte α-Naphthyl Butyrate Esterase Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352108
NACRES:
NA.47
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

Expiry date on the label.

Quality Segment

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

General description

Demonstration of lymphocyte staining patterns may indicate specific stages of maturation arrest associated with certain lymphoproliferative disorders.
This kit is intended for demonstration of nonspecific esterase in a subset of lymphocytes for determining maturation arrest. Kits 90A1 or 91A are intended for monocyte identification.


Kit Components Only

Product No.
Description

  • α-Naphthyl Butyrate Solution (1801) 50 mL

  • Citrate Solution (915) 50 mL

  • Methylene Blue Solution (1808) 50 mL

  • Pararosaniline Solution (1804) 15 mL

  • Phosphate Buffer (1805) 1 mL/vial

  • Sodium Nitrite (kit only) 10 tablets

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Ox. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

98.1 °F

flash_point_c

36.7 °C

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
181B-PART104061834410430
181B-1KT04061838755544
HMI0270-5NMOL04061831381696
HMI1966-5NMOL04061831377613