Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17804

Sigma-Aldrich

Isovitexin

≥98.0% (HPLC)

Synonym(s):

6-C-Glucosylapigenin, Homovitexin, Saponaretin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
66651
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

form

powder

color

white to light yellow

mp

220 -221  °C ((428 - 430 °F))

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)cc4

InChI

1S/C21H20O10/c22-7-14-17(26)19(28)20(29)21(31-14)16-11(25)6-13-15(18(16)27)10(24)5-12(30-13)8-1-3-9(23)4-2-8/h1-6,14,17,19-23,25-29H,7H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

MYXNWGACZJSMBT-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isovitexin can be used to study biochemicals found in plants and nutrition. Isovitexin has been used in a study to assess collagenase inhibitors from Viola yedoensis. Isovitexin has also been used in a study to investigate the inhibitory activity of Gentiana lutea extracts on the enzyme myeloperoxidase (MPO), as well as the antioxidant activity of these extracts and their correlation with the total polyphenol content.

Biochem/physiol Actions

Phytochemical found in medicinal herbs. Antioxidant.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 5

1 of 5

Kazuki Kanazawa et al.
Journal of agricultural and food chemistry, 60(17), 4359-4368 (2012-04-18)
It is desirable to increase the flavonoid contents of postharvest vegetables since flavonoids play a beneficial role in human health promotion. In the present study, we show that postharvest vegetables increasingly produced flavonoids when irradiated with light near the absorption
Branislav Nastasijević et al.
Journal of pharmaceutical and biomedical analysis, 66, 191-196 (2012-04-24)
The aim of this study was to investigate the inhibitory activity of Gentiana lutea extracts on the enzyme myeloperoxidase (MPO), as well as the antioxidant activity of these extracts and their correlation with the total polyphenol content. Extracts were prepared
He Li et al.
Food chemistry, 135(4), 2942-2946 (2012-09-18)
Mung bean soup (MBS) has been traditionally taken as a kind of health food in China. To learn the mechanisms underlying its health benefits, antioxidant capacities of the soup prepared with three cultivars of mung bean were measured. The highest
Poliane Folador et al.
Fitoterapia, 81(8), 1180-1187 (2010-08-04)
The antihyperglycemic effect and mechanism of action of extracts, fractions and compounds from Wilbrandia ebracteata was studied. The crude extract reduced the glycemia, increased glycogen content and serum insulin in hyperglycemic rats. Also, a significant effect was observed with the
Yujie Fu et al.
Journal of chromatography. A, 1139(2), 206-213 (2006-12-05)
Vitexin and isovitexin are a pair of isomeric compounds known as the major constituents in pigeonpea leaves and possess various pharmacological activities. In the present study, the preparative separation of vitexin and isovitexin with macroporous resins (Nankai Hecheng S &

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service