Skip to Content
MilliporeSigma
All Photos(2)

Documents

150-05A

Sigma-Aldrich

Human Skeletal Muscle Cells: HSkMC, adult

Synonym(s):

Human Muscle Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human skeletal muscle (normal limb)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

Skeletal muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

muscular dystrophy

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Human Skeletal Muscle Cells (HSkMC) are isolated from the skeletal muscle of limbs from adult or fetal donors. Human Skeletal Muscle Cells can undergo differentiation to exhibit actin and myosin myofilaments. HSkMC from Cell Applications, Inc. provide a useful model system to study many aspects of muscular function and disease.Skeletal Muscle Cells play an instrumental role in the glucose metabolism and diabetes, therefore select HSkMC lots undergo an additional set of characterization at Cell Applications, Inc. to demonstrate AMPK signaling and responsiveness to insulin stimulation.

Cell Line Origin

Muscle (Limb)

Application

lipid and glucose metabolism, exercise, insulin action, cell signaling

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 15 doublings

Subculture Routine

Please refer to the HSkMC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human skeletal muscle cells including thawing, subculturing and cryopreservation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service