Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12676

Sigma-Aldrich

Apigenin 7-O-neohesperidoside

≥99.0% (HPLC)

Synonym(s):

Rhoifolin, Rhoifoloside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥99.0% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to light yellow

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C=C(Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-8,10,18,20-30,32-36H,9H2,1H3/t10-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

RPMNUQRUHXIGHK-PYXJVEIZSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Hua Liu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 35(1), 56-61 (2012-06-28)
To investigate the impurities in naringin extracted from Citrus grandis 'Tomentosa'. High performance liquid chromatographies coupled with photodiode array and electrospray ionization mass spectrometry detectors (HPLC-PDA/ESI-MS/MS) were applied to investigate the impurities, and their structures were elucidated by spectral data
K Ishii et al.
Journal of chromatography. B, Biomedical applications, 655(2), 300-304 (1994-05-13)
A method for determining flavonoids in human plasma is presented for application to pharmacokinetic studies of two flavonoids, rhoifolin and daidzin. Isocratic reversed-phase high-performance liquid chromatography (HPLC) was used with genistin as an internal standard and solid-phase extraction using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service