Skip to Content
MilliporeSigma
All Photos(1)

Documents

12106C

Sigma-Aldrich

Fetal Bovine Serum

USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture

Synonym(s):

HI FBS, HI FCS, HI Fetal Bovine Sera, HI Fetal Bovine Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Premier

composition

hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Fetal Bovine Serum (FBS) provides many non-defined growth promoting and survival enhancing factors to cells in culture. FBS, heat inactivated may be used in place of regular FBS wherein the inactivation of the complement system is beneficial to cell growth or function. Heat-inactivated FBS is used to culture Leishmania sp.

Preparation Note

Prepared by heating at 56°C for 30 minutes.
Prepared by heating at 56°C for 30 minutes. Heating serum to 56°C for 30 minutes is used to inactivate the complement system for immunoassays. Heat activation also inactivates other undetermined inhibitors of cell growth in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Anitua et al.
Experimental eye research, 119, 27-34 (2013-12-19)
We have developed and characterized a new type of plasma rich in growth factors (PRGF) derived eye-drop therapy for patients suffering from autoimmune diseases. To determine the concentration of several growth factors, proteins, immunoglobulins and complement activity of the heat-inactivated
Nand Lal Sharma et al.
The American journal of tropical medicine and hygiene, 72(6), 819-824 (2005-06-21)
Localized cutaneous leishmaniasis (LCL) in India is due mostly to Leishmania tropica. It is mainly endemic in the deserts of Rajasthan. Recently, Himachal Pradesh has been identified as a new endemic focus for the disease. In the last few years
Li Zhou et al.
Genome biology, 9(7), R119-R119 (2008-07-31)
CD8+ NKT-like cells are naturally occurring but rare T cells that express both T cell and natural killer cell markers. These cells may play key roles in establishing tolerance to self-antigens; however, their mechanism of action and molecular profiles are
Ruth M Risueño et al.
PloS one, 3(3), e1747-e1747 (2008-03-06)
How the T cell antigen receptor (TCR) discriminates between molecularly related peptide/Major Histocompatibility Complex (pMHC) ligands and converts this information into different possible signaling outcomes is still not understood. One current model proposes that strong pMHC ligands, but not weak
Silvia Goldoni et al.
The American journal of pathology, 173(3), 844-855 (2008-08-09)
Decorin, a member of the small leucine-rich proteoglycan gene family, down-regulates members of the ErbB receptor tyrosine kinase family and attenuates their signaling, leading to growth inhibition. We investigated the effects of decorin on the growth of ErbB2-overexpressing mammary carcinoma

Protocols

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service