Skip to Content
MilliporeSigma

10021101

SerBob cell line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human prostate
Relevant disease(s):
cancer
Growth mode:
Adherent
Morphology:
Cobblestone
Receptors:
CD44, alpha2 integrin, alpha6 integrin, EGFR, FGFR1, LIFR
Technique(s):
cell culture | mammalian: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human prostate

Quality Segment

form

liquid

manufacturer/tradename

ECACC

packaging

tube of 5 μg 10021101-DNA-5UG, pkg of vial of cells 10021101-1VL

growth mode

Adherent

karyotype

4n (67 to 88)

morphology

Cobblestone

products

More differentiated than Bob with increased CD24, PAP, PSCA expression (prostate differentiation markers). 10% 3-D sphere formation in Matrigel. Migratory and invasive when compared to benign prostate tumour. More invasive than Bob.

receptors

CD44, alpha2 integrin, alpha6 integrin, EGFR, FGFR1, LIFR

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Prostate cancer research including drug testing.

Biochem/physiol Actions

First spontaneously immortalized prostate cancer cell line to be established from a trans-rectal needle biopsy (TRBP) of a patient with castration-resistant prostate cancer (CRPC). Bob is a novel pre-clinical model for functional studies in CRPC and especially for studying the CRPC "basal" phenotype. SerBob grows as a monolayer culture with limited differentiation. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.
Human prostate cancer spontaneously immortalized
STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 9,14
D16S539: 10,12
D5S818: 13
D7S820: 10,11
THO1: 9
TPOX: 8,10
vWA: 17

Packaging

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Preparation Note

Keratinocyte-SFM (Invitrogen, catalogue no 37010-022, supplied with prequalified human recombinant epidermal growth factor 1-53 (EGF 1-53), bovine pituitary extract (BPE) and glutamine) + 2ng/ml leukaemia inhibitory factor + 2ng/ml stem cell factor + 100ng/ml cholera toxin + 1ng/ml granulocyte macrophage colony stimulating factor.
Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 2-4 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. At confluence 5-7 x 104 cells/cm2 can be expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library