Skip to Content
MilliporeSigma

07294

O′-(Carboxymethyl)fluoresceinamide

BioReagent, suitable for fluorescence, ≥97.0% (HPCE)

Synonym(s):

[9-(2-Dimethylcarbamoylphenyl)-6-oxo-6H-xanthen-3-yloxy]acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H19NO6
CAS Number:
Molecular Weight:
417.41
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥97.0% (HPCE)

fluorescence

λex 458 nm; λem 520 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CN(C)C(=O)c1ccccc1C2=C3C=CC(=O)C=C3Oc4cc(OCC(O)=O)ccc24

InChI

1S/C24H19NO6/c1-25(2)24(29)17-6-4-3-5-16(17)23-18-9-7-14(26)11-20(18)31-21-12-15(8-10-19(21)23)30-13-22(27)28/h3-12H,13H2,1-2H3,(H,27,28)

InChI key

IBIKQEXCWKEQLF-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library