Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05288

Sigma-Aldrich

K-252a

from Nocardiopsis sp.

Synonym(s):

Protein kinase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21N3O5
CAS Number:
Molecular Weight:
467.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.85

biological source

Nocardiopsis sp.

form

powder

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

[H][C@]12C[C@](O)(C(=O)OC)[C@](C)(O1)n3c4ccccc4c5c6CNC(=O)c6c7c8ccccc8n2c7c35

InChI

1S/C27H21N3O5/c1-26-27(33,25(32)34-2)11-18(35-26)29-16-9-5-3-7-13(16)20-21-15(12-28-24(21)31)19-14-8-4-6-10-17(14)30(26)23(19)22(20)29/h3-10,18,33H,11-12H2,1-2H3,(H,28,31)/t18-,26+,27+/m1/s1

InChI key

KOZFSFOOLUUIGY-SOLYNIJKSA-N

Gene Information

human ... NTRK1(4914)
rat ... Prkca(24680)

Looking for similar products? Visit Product Comparison Guide

Application

K-252a is used to inhibit protein kinases involved in cell signaling processes, including CaM kinase; phosphorylase kinase, serine/threonine kinases, Trk. It is used to inhibit the actions of nerve growth factor (NGF) on PC1 2 cells and to inhibit smooth muscle myosin light chain kinase .

Biochem/physiol Actions

Potent inhibitor of various protein kinases including protein kinase A, C, and G. It is is reported to selectively inhibit the actions of nerve growth factor (NGF) on PC1 2 cells and has been shown to inhibit smooth muscle myosin light chain kinase .

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nakanishi et al.
The Journal of biological chemistry, 263(13), 6215-6219 (1988-05-05)
Effects of K-252a, (8R*, 9S*, 11S*)-(-)-9-hydroxy-9-methoxycarbonyl-8-methyl-2,3,9,10-tetrahydro-8, 11-epoxy-1H,8H,11H-2,7b,11a-triazadibenzo[a,g]cycloocta [cde]trinden-1-one, purified from the culture broth of Nocardiopsis sp., on the activity of myosin light chain kinase were investigated. 1) K-252a (1 x 10(-5) M) affected three characteristic properties of chicken gizzard myosin-B
S Koizumi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 8(2), 715-721 (1988-02-01)
K-252a, a kinase inhibitor isolated from the culture broth of Nocardiopsis sp., selectively inhibits the actions of nerve growth factor (NGF) on PC 12 cells. At a concentration of 200 nM, K-252a prevents neurite generation initiated by NGF, but not
Inhibition of actin-myosin-ATP interaction of smooth muscle native actomyosin is not closely associated with inhibition of its myosin light chain phosphorylation: studies with derivatives of antibiotic SF-2370.
K Kohama et al.
Progress in clinical and biological research, 315, 460-462 (1989-01-01)
A new antibiotic SF-2370 produced by Actinomadura.
M Sezaki et al.
The Journal of antibiotics, 38(10), 1437-1439 (1985-10-01)
Monique Rascher et al.
Biomolecules, 10(10) (2020-10-18)
Medicinal mushrooms of the genus Hericium are known to produce secondary metabolites with homeostatic properties for the central nervous system. We and others have recently demonstrated that among these metabolites cyathane diterpenoids and in particular erinacine C possess potent neurotrophin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service