Skip to Content
MilliporeSigma

02350

Esculin hydrate

≥98.0% (HPLC)

Synonym(s):

(-)-Esculin, 6,7-Dihydroxycoumarin 6-glucoside, Esculetin 6-β-D-glucoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H16O9 · xH2O
CAS Number:
Molecular Weight:
340.28 (anhydrous basis)
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D −41±2°, c = 5% in pyridine

impurities

≤10.5% water

solubility

methanol: 50 mg/mL, clear to very faintly turbid, yellow-green (fluoresc.)

SMILES string

O.OC[C@H]1O[C@@H](Oc2cc3C=CC(=O)Oc3cc2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H16O9.H2O/c16-5-10-12(19)13(20)14(21)15(24-10)23-9-3-6-1-2-11(18)22-8(6)4-7(9)17;/h1-4,10,12-17,19-21H,5H2;1H2/t10-,12-,13+,14-,15-;/m1./s1

InChI key

CQYPGSKIFJFVDQ-QWFKVUSTSA-N

Other Notes

Wavelength-shifter, 0,4 g/L improve counting efficiency by a factor of 1.3 when 32P-Cerenkov radiation is measured in aqueous samples


Still not finding the right product?

Explore all of our products under Esculin hydrate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library