Skip to Content
MilliporeSigma
All Photos(4)

Documents

675989

Sigma-Aldrich

Cyclopentyl methyl ether

greener alternative

contains 50 ppm BHT as inhibitor, ReagentPlus®, ≥99.90%

Synonym(s):

CPME

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99.90%

form

liquid

contains

50 ppm BHT as inhibitor

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤50 ppm H2O2

bp

106 °C/760 mmHg

mp

-140 °C

density

0.86 g/mL at 25 °C

greener alternative category

SMILES string

COC1CCCC1

InChI

1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

SKTCDJAMAYNROS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Cyclopentyl methyl ether (CPME) is a greener alternative solvent to many aprotic ether solvents such as tetrahydrofuran (THF), 2-methyl tetrahydrofuran (2-MeTHF), dioxane (carcinogenic), tert-Butyl methyl ether (MTBE), and 1,2-dimethoxyethane (DME) and thus has been enhanced for "Safer Solvents and Auxiliaries".  Click here  for more information.

Application

CPME is a greener alternative solvent and may be used is a broad range of applications such as:

Organic synthesis reactions containing the following reagents and conditions:

  1. Alkali agents
  2. Lewis’s acid-mediated reactions
  3. Organometallic
  4. Reduction and Oxidation
  5. Transition metal catalysts
  6. Azeotropic water removal



Extraction of compounds
Polymer synthesis
Crystallization
Surface coatings

Features and Benefits

  • Synthetic-based sustainable and safer alternative
  • Reduces the number of steps required for production
  • Reduces the amount of water that is required for washing
  • Eliminates the need for co-solvents for product recovery

    It has many properties which make it a safer and greener alternative:
  • Low peroxide formation
  • High boiling point
  • Clean organic-water phase separation
  • Reduced energy to recover

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

<37.4 °F - closed cup

Flash Point(C)

< 3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Replacement of dichloromethane within chromatographic purification: a guide to alternative solvents
MacMillan, D.S.; et al.
Green Chemistry (2012)
Cyclopentyl Methyl Ether as a New and Alternative Process Solvent
Watanabe, K.; Yamagiwa, N.; Torisawa, Y.
Organic Process Research & Development, 11 (2007)
Rikako Takahiro et al.
Biological & pharmaceutical bulletin, 38(1), 58-65 (2015-03-07)
The aim of this study was to characterize the kinetics of metabolite formation of the phosphodiesterase type-5 (PDE5) inhibitors sildenafil and tadalafil by CYP3A4, CYP3A5, and CYP3A7 isoforms. The formations of N-desmethyl sildenafil and desmethylene tadalafil were examined using CYP3A

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service