Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412236

Sigma-Aldrich

Octane

reagent grade, 98%

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696875
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

3.9 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

428 °F

expl. lim.

6.5 %

refractive index

n20/D 1.398 (lit.)

bp

125-127 °C (lit.)

mp

−57 °C (lit.)

density

0.703 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octane is an aliphatic hydrocarbon that is widely used as a solvent in paint, adhesives, and liquid process photocopiers.

Application

Octane can be used as a solvent in the:
  • Selective hydrogenation of furfural.
  • Metal-free oxidation of aromatic thiophenes to sulfones.
  • Ni-catalyzed β-alkylation of alcohols.
  • Enzyme-catalyzed epoxidation of unconjugated terminal alkenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comprehensive detailed chemical kinetic reaction mechanism for combustion of n-alkane hydrocarbons from n-octane to n-hexadecane.
Westbrook CK, et al.
Combustion and Flame, 156(1), 181-199 (2009)
Adsorption equilibrium and kinetics of dibenzothiophene from n-octane on bamboo charcoal.
Zhao D, et al.
Applied Surface Science, 254(10), 3242-3247 (2008)
Fernando E Maturi et al.
Materials (Basel, Switzerland), 12(6) (2019-03-25)
Mesoporous silica nanoparticles prepared by organic template-driven synthesis have been successfully explored as carriers of the drug-derivate green luminescent complex of terbium (III) with the nonsteroidal anti-inflammatory drug ketoprofen. The terbium (III) complex was synthesized by reacting ketoprofen sodium salt
Heat capacities and densities of liquid n-octane, n-nonane, n-decane, and n-hexadecane at temperatures from 318.15 K to 373.15 K and at pressures up to 10 MPa.
Banipal TS, et al.
The Journal of Chemical Thermodynamics, 23(10), 923-931 (1991)
Effective production of octane from biomass derivatives under mild conditions.
Xu W, et al.
ChemSusChem, 4(12), 1758-1761 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service