Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

310360

Sigma-Aldrich

Aluminum

ACS reagent, 99%, wire, wire diam. ~1.5 mm

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

99%

form

wire

autoignition temp.

1400 °F

reaction suitability

reagent type: catalyst
core: aluminum

resistivity

2.6548 μΩ-cm

wire diam.

~1.5 mm

impurities

≤0.001% N compounds
≤0.05% insol. dil. HCl

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

cation traces

Cu: ≤0.02%
Fe: ≤0.1%
Mn: ≤0.002%
Si: ≤0.1%
Ti: ≤0.03%

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Application


  • A recyclable luminescent MOF sensor for on-site detection of insecticide dinotefuran and anti-Parkinson′s drug entacapone in various environmental and biological specimens.: This study highlights the development of a metal-organic framework (MOF) sensor that can detect traces of dinotefuran and entacapone, demonstrating potential applications in environmental monitoring and healthcare diagnostics (Mir NUD et al., 2024).

  • A lithium-aluminium heterobimetallic dimetallocene.: This paper introduces a novel bimetallic compound combining lithium and aluminium, offering insights into their cooperative effects in chemical synthesis and potential applications in material science (Bischoff IA et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate
Caitlin Carney et al.
Journal of the American Academy of Dermatology, 69(4), 578-582 (2013-07-17)
Laser treatment has emerged as a novel treatment modality for onychomycosis. We sought to determine thermal response and optical effects of a submillisecond neodymium:yttrium-aluminum-garnet (Nd:YAG) 1064-nm laser on common fungal nail pathogens, and the clinical efficacy and safety of the
Jesper C Holst et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8819-8823 (2013-05-15)
Refractory inclusions [calcium-aluminum-rich inclusions, (CAIs)] represent the oldest Solar System solids and provide information regarding the formation of the Sun and its protoplanetary disk. CAIs contain evidence of now extinct short-lived radioisotopes (e.g., (26)Al, (41)Ca, and (182)Hf) synthesized in one
Arjun Joshua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), 9633-9638 (2013-05-28)
Controlling the coupling between localized spins and itinerant electrons can lead to exotic magnetic states. A novel system featuring local magnetic moments and extended 2D electrons is the interface between LaAlO3 and SrTiO3. The magnetism of the interface, however, was
Mutlu Ozcan et al.
The journal of adhesive dentistry, 15(3), 211-214 (2013-05-24)
To evaluate the effect of nozzle distance, nozzle angle, and deposition duration on the silica content attained on zirconia by air abrasion. Disk-shaped zirconia (LAVA, 3M ESPE) (diameter: 10 mm, thickness: 2 mm) specimens (N = 54) were obtained. They

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service