Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

295876

Sigma-Aldrich

Formamide

spectrophotometric grade, ≥99%

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.07

grade

spectrophotometric grade

Quality Level

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥99%

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 %

technique(s)

UV/Vis spectroscopy: suitable
hybridization: suitable

impurities

<0.030% water

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 275 nm Amax: 1.00
λ: 280 nm Amax: 0.20
λ: 300 nm Amax: 0.05
λ: 360 nm Amax: 0.02
λ: 400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip Thomas et al.
Mutagenesis, 23(1), 57-65 (2007-12-01)
Alzheimer's disease (AD) is a premature ageing syndrome characterized by cognitive impairment arising from neuropathological changes occurring within specific areas of the brain. We report a 1.5-fold increase in trisomy 21 (P < 0.001) and a 1.2-fold increase in trisomy
J Casey et al.
Nucleic acids research, 4(5), 1539-1552 (1977-01-01)
The thermal stabilities of RNA:DNA hybrids are substantially greater than those of DNA:DNA duplexes in aqueous electrolyte solutions containing high concentrations of formamide. Association rates to form DNA:DNA duplexes and DNA:RNA hybrids have been measured in these solvents. There is
Microwave spectrum and molecular structure of formamide.
Costain CC and Dowling JM.
J. Chem. Phys. , 32(1), 158-165 (1960)
Solubilization in formamide protects RNA from degradation.
Chomczynski P.
Nucleic Acids Research, 20(14), 3791-3791 (1992)
The formamide method for the extraction of polysaccharides from haemolytic streptococci.
Fuller AT.
British Journal of Experimental Pathology, 32(1), 130-130 (1960)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service