Skip to Content
MilliporeSigma
All Photos(3)

Documents

255521

Sigma-Aldrich

Strontium chloride hexahydrate

ACS reagent, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCl2 · 6H2O
CAS Number:
Molecular Weight:
266.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

99%
99.0-103.0% (ACS specification)

form

chunks
crystalline powder
powder, crystals or granules

impurities

≤0.005% insolubles

pH

5.0-7.0 (25 °C, 5%)

mp

115 °C (lit.)

anion traces

sulfate (SO42-): ≤0.001%

cation traces

Ba: ≤0.05%
Ca: ≤0.05%
Fe: ≤5 ppm
Mg: ≤2 ppm
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.O.O.O.O.Cl[Sr]Cl

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of cadmium sulfide core photocatalytic nanoparticles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahang, H., et al.
Mat. Res. Bul., 38, 1033-1033 (2003)
Davood Azarifar et al.
Molecules (Basel, Switzerland), 15(3), 1433-1441 (2010-03-26)
SrCl2 x 6 H2O has been shown to act as an efficient catalyst for the conversion of aldehydes or ketones into the corresponding gem-dihydroperoxides (DHPs) by treatment with aqueous H2O2 (30%) in acetonitrile. The reactions proceed under mild and neutral
Unstable intermediates. Part LX. HO2 radical in γ-irradiated strontium chloride hexahydrate.
Catton RC and Symons MCR.
J. Chem. Soc. Sect. A, 1393-1395 (1969)
Ke Zhang et al.
Molecular cell, 79(2), 234-250 (2020-06-25)
Somatic cell nuclear transfer (SCNT) can reprogram a somatic nucleus to a totipotent state. However, the re-organization of 3D chromatin structure in this process remains poorly understood. Using low-input Hi-C, we revealed that, during SCNT, the transferred nucleus first enters
David L Bryce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(17), 4786-4796 (2007-03-27)
A series of alkaline earth chloride hydrates has been studied by solid-state (35/37)Cl NMR spectroscopy in order to characterize the chlorine electric field gradient (EFG) and chemical shift (CS) tensors and to relate these observables to the structure around the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service