Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

244023

Sigma-Aldrich

Potassium hexacyanoferrate(III)

ACS reagent, ≥99.0%

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Assay

≥99.0%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.005% insolubles

pH

6-9 (25 °C, 329 g/L)

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.05%

storage temp.

room temp

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hexacyanoferrate(III) (Potassium ferricyanide) can be prepared by the anodic oxidation of K4[Fe(CN)6].

Application

Potassium hexacyanoferrate(III) [Potassium ferricyanide] has been used as an oxidant in the preparation of diethidium and di-mito-ethidium. It reacts with cobalt chloride hexahydrate to afford CoHCF (Cobalt hexacyanoferrate) nanoparticles.
It may be employed as an oxidizing agent during the fabrication of glucose conductometric biosensor.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 291-291 (1994)
Macro-micro relationship in nanostructured functional
Composites
Zanotto A, et al.
Express Polymer Letters, 6(5), 410-416 (2012)
Glucose conductometric biosensor with potassium hexacyanoferrate (III) as an oxidizing agent.
Dzyadevich SV, et al.
Analytica Chimica Acta, 374(1), 11-18 (1998)
Thorsten Litzenburger et al.
Clinical science (London, England : 1979), 134(7), 921-939 (2020-04-03)
Maternal obesity determines obesity and metabolic diseases in the offspring. The white adipose tissue (WAT) orchestrates metabolic pathways, and its dysfunction contributes to metabolic disorders in a sex-dependent manner. Here, we tested if sex differences influence the molecular mechanisms of
Jacek Zielonka et al.
Free radical biology & medicine, 46(3), 329-338 (2008-11-26)
Methods for the detection and quantitation of hydroethidine (HE) and its oxidation products by HPLC analysis are described. Synthetic methods for preparation of authentic standards (2-hydroxyethidium and diethidium) are provided. Potential applications of the HPLC methods to chemical and biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service