Skip to Content
MilliporeSigma
All Photos(2)

Documents

226823

Sigma-Aldrich

Silver iodide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgI
CAS Number:
Molecular Weight:
234.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Assay

99%

form

powder

density

5.68 g/mL at 25 °C (lit.)

SMILES string

[Ag]

InChI

1S/Ag.HI/h;1H/q+1;/p-1

InChI key

MSFPLIAKTHOCQP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Negative Differential Resistance (NDR) Memristors: Silver iodide is used in the development of bulk memristors exhibiting negative differential resistance (NDR). These memristors are essential for creating non-volatile memory devices and neuromorphic computing systems. The compositional tuning of AgI memristors has been shown to enhance their performance, making them vital components in advanced electronic applications (Naik and Rabinal, New Journal of Chemistry, 2021).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereochemical systematics of metal clusters. Crystallographic evidence for a new cubane. dblharw. chair isomerism in tetrameric triphenylphosphine silver iodide,(Ph3P) 4Ag4I4.
Teo BK and Calabrese JC.
Inorganic Chemistry, 15(10), 2474-2486 (1976)
Behavior of the low-frequency conductivity of silver iodide nanocomposites in the superionic phase transition region.
Vergent'ev TV, et al.
Physics of the Solid State, 55(1), 175-180 (2013)
Formation, structure, and polymorphism of novel lowest-dimensional AgI nanoaggregates by encapsulation in carbon nanotubes.
Matteo Baldoni et al.
Small (Weinheim an der Bergstrasse, Germany), 3(10), 1730-1734 (2007-09-13)
Xunsi Wang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 586-589 (2011-12-06)
A serious of tellurium based chalcogenide glass were prepared and investigated. As it being transparent nearly up to 25 μm and strong anti-hydrability, it becomes an optimized material for far-infrared application. Here, AgI was incorporated into the glasses acting as
Biao Xu et al.
Small (Weinheim an der Bergstrasse, Germany), 7(24), 3439-3444 (2011-11-01)
AgI quantum dot (QD)-based room-temperature fast ionic conductors are prepared via an aqueous route. New phase behavior and good performance are found in this material. This is a prototype of quantum dot-ionics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service