Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

213349

Sigma-Aldrich

Ammonium bromide

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Br
CAS Number:
Molecular Weight:
97.94
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050407
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.0%
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Quality Level

vapor pressure

1 mmHg ( 198.3 °C)

Assay

≥99.0%

form

powder or crystals

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

4.5-6.0 (25 °C, 5%)

mp

452 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

anion traces

bromate (BrO3-): ≤0.002%
chloride (Cl-): ≤0.2%
iodide (I-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Fe: ≤5 ppm
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

N.Br[H]

InChI

1S/BrH.H3N/h1H;1H3

InChI key

SWLVFNYSXGMGBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Laser excited Raman spectral study reveals that 1?0 torsional transitions of the NH4+ ion in phase III of NH4Br were observed at 331cm-1. Neutron diffraction studies have been conducted to evaluate the P-T phase diagram of NH4Br and its deuterated form (ND3Br).

Application

Ammonium bromide may be used in the preparation of organic ammonium tribromides. It may also be used as a reagent during the transformation of benzyl derivatives of halides, amines, alcohols, and aldehydes into respective nitriles via oxidation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Repr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neutron-diffraction measurements on the P-T phase diagram of ammonium bromide.
Press W, et al.
Physical Review. B, Condensed Matter and Materials Physics, 14(5), 1983-1983 (1976)
Direct oxidative conversion of benzylhalides,-amines,-alcohols, and arylaldehydes to nitriles with trans-3, 5-dihydroperoxy-3, 5-dimethyl-1, 2-dioxolane activated by NH4Br.
Azarifar D and Najminejad Z.
Journal of the Iranian Chemical Society, 12(1), 107-111 (2015)
Veerababurao Kavala et al.
The Journal of organic chemistry, 70(11), 4267-4271 (2005-05-21)
1,2-Dipyridiniumditribromide-ethane (DPTBE) has been synthesized and explored as a new efficient brominating agent. The crystalline ditribromide reagent is stable for months and acts as a safe source of bromine requiring just 0.5 equiv for complete bromination. It has high active
Low-Frequency Vibrations in Ammonium Iodide and Ammonium Bromide.
Durig JR and Antion DJ.
J. Chem. Phys. , 51(9), 3639-3647 (1969)
Emilia Fisicaro et al.
The journal of physical chemistry. B, 109(5), 1744-1749 (2006-07-21)
The apparent and partial molar enthalpies, apparent molar volumes, and adiabatic compressibilities at 298 K of the aqueous solutions of the cationic gemini surfactants propanediyl-alpha,omega-bis(octyldimethylammonium bromide) (8-3-8) and propanediyl-alpha,omega-bis(dodecyldimethylammonium bromide) (12-3-12) have been measured as a function of concentration. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service