Skip to Content
MilliporeSigma
All Photos(1)

Documents

12648

Sigma-Aldrich

Potassium nitrate

puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Assay

99.0-100.5%

form

powder or crystals

quality

meets analytical specification of Ph Eur, BP, USP, FCC, E252

impurities

acidity or alkalinity, complies
reducing matter, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.005% ammonium (NH4)

loss

≤0.5% loss on drying, 105 °C

pH

5-8 (20 °C, 5%)

mp

334 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
nitrite (NO2-): ≤5 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤50 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Na: ≤1000 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium nitrate is colorless potassium salt having rhombic crystals. It is an oxidizing agent. In nature it is found as saltpeter. It can be prepared by reacting potassium carbonate/ hydroxide with nitric acid or by the reaction of sodium nitrate with potassium chloride.1 It can be prepared by combining ammonium nitrate and potassium hydroxide under hot conditions.
Potassium nitrate can find applications as an oxidizer, which can be prepared by combining ammonium nitrate and potassium hydroxide under hot conditions.

Application

Potassium nitrate may be employed for the following studies:
  • Preparation of gun powder, ceramics, glass, dyeing and printing.1
  • As oxidizing agent in the fireworks.1
  • Preparation of potassium nitrite.1
  • As desensitizing agent for tooth hypersensitivity.
  • As phase change material for the preparation of nanofluids with phase change behavior.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosanke.L.K, et al. et al.
Encyclopedic Dictionary of Pyrotechnics: (and Related Subjects) (2012)
A superior desensitizer--potassium nitrate.
M Hodosh
Journal of the American Dental Association (1939), 88(4), 831-832 (1974-04-01)
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Mila Ivanova Luleva et al.
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service