Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12353

Sigma-Aldrich

Iron(II) sulfate hydrate

puriss., meets analytical specification of Ph. Eur., BP, exsiccated, 86.0-89.0% FeSO4 basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · aq
CAS Number:
Molecular Weight:
151.91 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

Assay

86.0-89.0% FeSO4 basis

form

solid

quality

exsiccated
meets analytical specification of Ph. Eur., BP

shelf life

Shelf life is limited

reaction suitability

core: iron
reagent type: catalyst

impurities

residual solvents, complies
≤0.005% heavy metals (as Pb)
≤0.05% insoluble matter

pH

3.0-4.0 (20 °C, 5%)

anion traces

chloride (Cl-): ≤300 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤200 mg/kg
Cr: ≤100 mg/kg
Cu: ≤20 mg/kg
Fe3+: ≤4000 mg/kg
Hg: ≤1 mg/kg
K: ≤100 mg/kg
Mg: ≤100 mg/kg
Mn: ≤1000 mg/kg
Na: ≤200 mg/kg
Ni: ≤100 mg/kg
Pb: ≤2 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

XBDUTCVQJHJTQZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

bottled under inert gas

Caution

tends to oxidation by air

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service