Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12056

Sigma-Aldrich

Calcium sulfate dihydrate

puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4 · 2H2O
CAS Number:
Molecular Weight:
172.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.0-101.0% (based on anhydrous substance)
form:
powder
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

99.0-101.0% (based on anhydrous substance)

form

powder

quality

meets analytical specification of NF, E 516

impurities

residual solvents, in accordance
≤0.001% heavy metals (as Pb)

loss

19.0-23.0% loss on drying, 250 °C

anion traces

chloride (Cl-): ≤100 mg/kg
fluoride (F-): ≤30 mg/kg

cation traces

As: ≤3 mg/kg
Fe: ≤50 mg/kg
Hg: ≤10 mg/kg
Pb: ≤5 mg/kg
Se: ≤30 mg/kg

SMILES string

O.O.[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

PASHVRUKOFIRIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium sulfate dihydrate (gypsum) scale formation on heat exchanger surfaces: The influence of scale inhibitors.
Amjad Z.
Journal of Colloid and Interface Science, 123(2), 523-536 (1988)
K N Lewis et al.
Journal of materials science. Materials in medicine, 17(6), 531-537 (2006-05-13)
Calcium sulfate (CS) is one of the oldest bone graft materials still in use. Its main limitations are poor handling characteristics, poor mechanical properties, and a resorption rate that is too fast for some applications. The present study investigated the
The crystallization of calcium sulfate dihydrate.
Smith BR and Sweett F.
Journal of Colloid and Interface Science, 37(3), 612-618 (1971)
Kathryn A Dooley et al.
The Analyst, 138(17), 4838-4848 (2013-06-27)
In situ chemical imaging techniques are being developed to provide information on the spatial distribution of artists' pigments used in polychrome works of art such as paintings. The new methods include reflectance imaging spectroscopy and X-ray fluorescence mapping. Results from
Massimo Peruffo et al.
Physical chemistry chemical physics : PCCP, 15(6), 1956-1965 (2012-12-22)
Dissolution processes at single crystal surfaces often involve the initial formation and expansion of localized, characteristic (faceted) etch-pits at defects, in an otherwise comparatively unreactive surface. Using natural gypsum single crystal as an example, a simple but powerful morphological analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service