Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11607

Sigma-Aldrich

Boric acid

puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

2.6 mmHg ( 20 °C)

grade

puriss.

Assay

99.5-100.5%

form

powder

quality

meets analytical specification of Ph. Eur., BP, NF

impurities

organic substances, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)

loss

≤0.5% loss on drying, 5 h (on silica gel)

pH

3.8-4.8 (20 °C, 3.3%)

mp

160 °C (dec.) (lit.)

solubility

ethanol: in accordance

density

1.440 g/cm3

anion traces

sulfate (SO42-): ≤400 mg/kg

cation traces

As: ≤5 mg/kg
Fe: ≤5 mg/kg

suitability

complies for appearance of solution

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catanzaro EJ.
Boric Acid: Isotopic and Assay Standard Reference Materials, 1-1 (1970)
One-pot synthesis of 3, 4-dihydropyrimidin-2(1H)-ones using boric acid as catalyst.
Tu S, et al.
Tetrahedron Letters, 44(32), 6153-6155 (2003)
L Romano et al.
The Journal of antimicrobial chemotherapy, 55(1), 110-114 (2004-12-03)
There is very little information, to date, on the antifungal activity of bergamot oil. In this study, we investigated the in vitro activity of three bergamot oils (natural essence, furocoumarin-free extract and distilled extract) against clinically relevant Candida species. We
L Charlet Bhami et al.
Journal of vector borne diseases, 52(2), 147-152 (2015-06-30)
The use of low concentrations of boric acid as a potential and effective control agent for the eggs and immature stages of Aedes aegypti L. and Aedes albopictus Skuse (Diptera: Culicidae) is found to be safe and effective as compared
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service