Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002004

Arsenic solution

European Pharmacopoeia (EP) Reference Standard, 1.02 mg/mL (dissolved in aqueous nitric acid)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

arsenic

manufacturer/tradename

EDQM

concentration

1.02 mg/mL (dissolved in aqueous nitric acid)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia

Application

Arsenic solution EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Respiratory organs,Cardio-vascular system,Gastro-intestinal system,Central nervous system,Nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service